missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ DMEM/Ham's F-12 50/50 Mix, Powder

Used for culture of variety of cell types under low serum conditions. Corning™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.

$96.17 - $101.15

Specifications

Storage Requirements 2° to 8°C
Concentration 1X
Format Powder
Product Type DMEM/Ham's F-12 50/50 Mix
Quantity 1 x 10L
View More Specs

Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
MT90091PB View Documents Corning™
90091PB
Each for $101.15
MT90090PB View Documents Corning™
90090PB
Each for $96.17
Description

Description

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation is without sodium bicarbonate, L-glutamine and phenol red
Specifications

Specifications

2° to 8°C
Powder
1 x 10L
1X
DMEM/Ham's F-12 50/50 Mix