Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Supplier: Corning™ 9120048
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
50 L | |
EVA | |
1/Pk. | |
Each |
Heat sealable flexible tubing | |
1/Ea. | |
Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i |