Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Phosphate Buffered Saline (1X)


With low endotoxin content

Manufacturer: corning™  21040CV

 View more versions of this product

Catalog No. MT21040CV

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Formulation Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L
Format Liquid
Storage Requirements 2° to 30°C
pH 7.3 to 7.5
Quantity 6 × 500mL
Concentration 1X