Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X


With low endotoxin content

Manufacturer: corning™  21030CM

 View more versions of this product

Catalog No. MT21030CM

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage Requirements 2° to 30°C
Formulation Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L
Concentration 1X
Without Additives N/A
Format Liquid
With Additives Calcium and Magnesium
pH 6.9 to 7.1
Quantity 6 × 1000mL