Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X


With low endotoxin content

Manufacturer: corning™  21031CV

 View more versions of this product

Catalog No. MT21031CV

  • / Case of 6 Each
 

Description & Specifications

Specifications

Formulation Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L
pH 7.3 to 7.5
Quantity 6 × 500mL
Without Additives Calcium and Magnesium
Storage Requirements 2° to 30°C
Concentration 1X
Format Liquid