Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X


With low endotoxin content

Manufacturer: corning™  21031LX

 View more versions of this product

Catalog No. MT-21-031-LX

  • / Each
 

Description & Specifications

Specifications

Quantity 1 × 10,000mL
Quantity 1 × 10,000mL
Formulation Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L
Concentration 1X
Storage Requirements 2° to 30°C
Without Additives Calcium and Magnesium
Concentration 1X
pH 7.3 to 7.5
Format Liquid