Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 10X


With low endotoxin content

Manufacturer: corning™  20031CV

 View more versions of this product

Catalog No. MT20031CV

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage Requirements 2° to 30°C
Formulation Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2g/L, Sodium Chloride 80g/L, Disodium Phosphate 11.50g/L
Quantity 6 × 500mL
Concentration 10X
Solution Type Dulbecco's Phosphate-Buffered Saline without Calcium and Magnesium
Format Liquid
pH 6 to 7