Please login to your online account to display your discounted pricing

Corning™ cellgro™ Ampicillin, Sodium Salt


Packaged in plastic bottles

Manufacturer: corning™  61238RH

 View more versions of this product

Catalog No. MT61238RH

  • / Each
 

Description & Specifications

Specifications

Storage 2° to 8°C
Storage Requirements 2° to 8°C
Packaging Plastic Bottle
Quantity 10g
Solution Type Ampicillin, Sodium Salt, Powder
Format Powder