Please login to your online account to display your discounted pricing

Corning™ cellgro™ Ampicillin, Sodium Salt

Packaged in plastic bottles

$152.90 - $513.74

Specifications

Format Powder
Packaging Type Plastic Bottle
Solution Type Ampicillin, Sodium Salt, Powder
Storage 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MT61238RM

 
corning™
61238RM
100g Each for $513.74

MT61238RH

 
corning™
61238RH
10g Each for $152.90
Description & Specifications

Specifications

Format Powder
Packaging Type Plastic Bottle
Solution Type Ampicillin, Sodium Salt, Powder
Storage 2° to 8°C