missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ CellBIND™ Surface Cell Culture Flasks

Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Manufacturer:  Corning™ C3291

 View more versions of this product

Catalog No. 05-539-103



Description

Description

 

  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested

 

 

  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System

 

Specifications

Specifications

7.1 oz.
HDPE
150 cm2
Polystyrene
Yes
Canted
Sterile
13 to 45 mL
Orange
Vent
210 mL
For optimal cell attachment
Lot numbers for ease in traceability
Rectangular
Corning Cellbind
Documents