Please login to your online account to display your discounted pricing

Corning™ CellBIND™ Surface Cell Culture Flasks

Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Manufacturer: corning™  3298

 View all options for this product

Catalog No. 08-757-212


Description & Specifications


Capacity (English) 8.45 oz.
Capacity (Metric) 250mL
Culture Area 175cm2
Closure Type Phenolic style
Surface Coating Corning™ Cellbind™
Barcode No
Material Polystyrene
Neck Style Angled
Volume (Metric) Working 35 to 52.5mL
Closure Color Orange
Closure Material HDPE
For Use With (Application) For optimal cell attachment
Graduated Yes
Shape Rectangular
Includes Lot numbers for ease in traceability
Sterile Yes
Capacity 8.45 oz., 250mL

  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested

  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System