Please login to your online account to display your discounted pricing

Corning™ CellBIND™ Surface Cell Culture Flasks


Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Manufacturer: corning™  C431328

 View more versions of this product

Catalog No. 08-757-211

  • / Case of 84 Each
 

Description & Specifications

Specifications

Closure Color Orange
Closure Material HDPE
For Use With (Application) For optimal cell attachment
Graduated Yes
Includes Lot numbers for ease in traceability
Shape Rectangular
Sterile Yes
Neck Style Angled
Volume (Metric) Working 35 to 52.5mL
Capacity (English) 8.45 oz.
Capacity (Metric) 250mL
Culture Area 175cm2
Closure Type Vented
Surface Coating Corning™ Cellbind™
Barcode Yes
Material Polystyrene
Capacity 8.45 oz., 250mL

  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested

  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System