missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ CellBIND™ Surface Culture Dishes

Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.

Manufacturer:  Corning™ C3296

 View more versions of this product

Catalog No. 07-202-516



Description

Description

  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
  • Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
  • 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
  • 245 mm square dishes offer 500 cm2 growth surface
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Manufactured from optically clear virgin polystyrene
  • Sterile
  • Nonpyrogenic


Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.

Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Specifications

Specifications

Clear
20 mm
Clear
Yes
100 mm
Polystyrene
Cell Culture Petri Dish
16.5 mL (Well)
40/Cs.
Round
Round
Polystyrene
55 cm2
Sterile
CellBIND
Yes
Lid
5/Pk.
SDS
Documents