Please login to your online account to display your discounted pricing

Corning™ Cell-Tak Cell and Tissue Adhesive


Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Manufacturer: corning™  C354242

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. CB40242

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Concentration 1 to 5µg/cm2 of growth surface depending on cell type
Formulation As liquid in 5% acetic acid
Description Molecular weight: 110 to 114kd
Storage Requirements Stable for at least three months at 2° to 8°C. Do not freeze.
Quantity 10mg
Source Polyphenolic Proteins secreted by Mytilus edulis
Product Type Cell and tissue adhesive

  • Polyphenolic proteins
  • Used as coating on substrate to immobilize cells or tissue
  • Coating is transparent and stable for 10 to 14 days at 2° to 8°C
  • Can simplify manipulation of biological samples in number of common in vitro techniques
  • Establishment of primary cultures
  • In situ hybridization
  • Immunoassays
  • Microinjection
  • Immunohistochemistry