Please login to your online account to display your discounted pricing

Corning™ Cell Expansion Bags


features a single-web polyolefin film with very good clarity when filled for viewing cultures, suited for scalable expansion

Manufacturer: corning™  9120084

 View more versions of this product

Catalog No. MT9120084

  • / Each
 

Description & Specifications

Specifications

For Use With Expansion and culture of non-adherent cells, ideal for basic research biopharmaceutical applications
Certifications/Compliance ANSI, AMI, ISO 11137
Quantity Each
Film Description Single web polyolefin gas permeable film with good clarity
Sterilization Method Radiation
Tubing Style Integrated
Fill Volume 190mL
Includes Reusable sampling valve
Product Type Cell Expansion Bag
Capacity (Metric) 500L
Port Detail Port 1- two leads with female luers and small pinch clamps, port 2 -dockable fill line with sampling valve and small pinch clamp
Storage Requirements -20° to 40°C