Please login to your online account to display your discounted pricing

Corning™ Cell Expansion Bags


features a single-web polyolefin film with very good clarity when filled for viewing cultures, suited for scalable expansion

Manufacturer: corning™  9120086

 View more versions of this product

Catalog No. MT9120086

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20° to 40°C
Product Type Cell Expansion Bag
Certifications/Compliance ANSI, AMI, ISO 11137
For Use With (Application) Expansion and culture of non-adherent cells, ideal for basic research biopharmaceutical applications
Tubing Style Integrated
Film Description Single web polyolefin gas permeable film with good clarity
Quantity Each
Fill Volume 633mL
Sterilization Method Radiation
Port Detail Port 1- two leads with female luers and small pinch clamps, port 2 -dockable fill line with sampling valve and small pinch clamp
Capacity (Metric) 3L
Includes Reusable sampling valve