Please login to your online account to display your discounted pricing

Corning™ Calcein AM Fluorescent Dye


Reliable and consistent dye used for the detection of live cells and key functional activities in variety of cell-based assays

Manufacturer: corning™  C354217

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

 Disclaimers

For research use only.

Catalog No. 08-774-501

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) BD Falcon™ FluoroBlok™ Cell culture inserts and insert systems to label cells
Quantity 1mg
Storage Requirements -20°C