Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Manufacturer:  Corning™ C999818
 View more versions of this product
Catalog No. 02-883-8E
Description
Description
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
  • Glued-in rubber lining firmly bonded to an interface of PTFE
  • Autoclavable and reusable
Specifications
Specifications
Yes
Black
Screw Cap
18-415
Phenolic
PTFE, Rubber
17.5mm
PTFE, Rubber
Documents