Please login to your online account to display your discounted pricing
Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Manufacturer: corning™  C999815
 View more versions of this product
Catalog No. 02-883-8D
  • / Case of 288 Each
Description & Specifications
Specifications
Product Type Screw Cap
Tip Material PTFE, Rubber
Thread Size 15-415
Liner Material PTFE, Rubber
Material Phenolic
Color Black
Height (Metric) 16mm
Autoclavable Yes
  • Glued-in rubber lining firmly bonded to an interface of PTFE
  • Autoclavable and reusable