Please login to your online account to display your discounted pricing
Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Manufacturer: corning™  C9998241
 View more versions of this product
Catalog No. 02-883-8B
  • / Case of 144 Each
Description & Specifications
Specifications
Liner Material PTFE, Rubber
Height (Metric) 18.5mm
Material Phenolic
Thread Size 24-410
Autoclavable Yes
Product Type Screw Cap
Tip Material PTFE, Rubber
Color Black
  • Glued-in rubber lining firmly bonded to an interface of PTFE
  • Autoclavable and reusable