Please login to your online account to display your discounted pricing
Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Manufacturer: corning™  C999824
 View more versions of this product
Catalog No. 02-883-8A
  • / Case of 12 Each
Description & Specifications
Specifications
Color Black
Liner Material PTFE, Rubber
Product Type Screw Cap
Height (Metric) 12.5mm
Thread Size 24-400
Tip Material PTFE, Rubber
Autoclavable Yes
Material Phenolic
  • Glued-in rubber lining firmly bonded to an interface of PTFE
  • Autoclavable and reusable