Please login to your online account to display your discounted pricing
Corning™ Black Phenolic Screw Caps
Resist effects of temperature and steam in autoclaving
Manufacturer: corning™  C999940
 View more versions of this product
Catalog No. 02-883-9D
  • / Case of 72 Each
Description & Specifications
Specifications
Autoclavable Yes
Neck Finish 40-400
Height (Metric) With Closure 12.5 mm
Material Phenolic
Color Black
Closure Liner Rubber
No. per Case 72 Cs.