Please login to your online account to display your discounted pricing
Corning™ BioCoat™ Tumor Invasion Multiwell Plate
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Manufacturer: corning™  C354167
 View more versions of this product
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog No. 08-772-61
  • / Each
Description & Specifications
Specifications
Quantity 1
No. of Wells 96
Pore Size 8μm
Type Multiwell Plate
Storage Requirements 20°C
Culture Area 0.08cm2
Material PET