Corning™ BioCoat™ Tumor Invasion Multiwell Plate
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Manufacturer:  Corning™ C354165
 View more versions of this product
Catalog No. 08-774-380
Description
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running cell migration and invasion studies.
Specifications
Specifications
24
PET
Multiwell Plate
1/Pk.
Matrigel™ Matrix
0.3 cm2
8 μm
1
1/Ea.