Please login to your online account to display your discounted pricing
Corning™ BioCoat™ TC-treated Multiwell Variety Packs
Choose from different groups of cellware conveniently packaged together in popular combinations
$436.37 - $438.65
Specifications
Storage Requirements 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
08-774-132
corning™
C354417
Cellware Variety Pack Case of 5 Each for $436.37
08-774-133
corning™
C354431
Cellware Variety Pack Pack of 5 Each for $438.65
Description & Specifications
Specifications
Storage Requirements 2° to 8°C
  • Selections include different combinations of extracellular matrix proteins and attachment factors
Multiwell Plate Variety Packs
  • 6-well
  • Pack I contains five plates (clear, round well, flat-bottom, TC-treated, with 9.6cm2 cell growth area)
  • Coated with one of the following: Collagen I, Collagen IV, Fibronectin, Laminin, and Poly-D-Lysine
  • Lids (Fisher P/N 08-774-133 only)