Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-L-Ornithine/Laminin Multiwell Plate

Substrate combination enhances adhesion of neurons to glass and plastic surfaces

$473.54 - $498.79

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Culture Area Format Type Price Quantity    

08-772-67

 
corning™
C354659
2cm2 24-well 24-Well Poly-L-Ornithine/Laminin Multiwell Microplate Each for $483.77

08-772-66

 
corning™
C354658
9.6cm2 6-well 6-Well Poly-L-Ornithine/Laminin Multiwell Microplate Case of 5 Each for $473.54

08-772-68

 
corning™
C354657
0.32cm2 96-well 96-Well Poly-L-Ornithine/Laminin Multiwell Microplate Each for $498.79
Description & Specifications

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
Shelf Life 3 Months
Source Poly-L-Ornithine and Laminin
Storage Requirements 2° to 8°C
Surface Coating ECM/Attachment Factors
Well Design Round

  • Each lot is tested for the ability to promote neurite outgrowth with rat glioma/mouse neuroblastoma cells
  • Tested and found negative for the presence of bacteria and fungi