Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-L-Lysine Multiwell Plate


Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells

Manufacturer: corning™  C356516

 View more versions of this product

Catalog No. 08-772-93

  • / Case of 50 Each
 

Description & Specifications

Specifications

Lid Yes
Source Poly-L-Lysine
Quantity 5/pk.; 50/cs.
Sterile No
Type 96-Well Poly-L-Lysine Multiwell Microplate
Storage Requirements 4° to 30°C
Well Color Clear
Format 96-well
Culture Area 0.32cm2
Surface Coating ECM/Attachment Factors
Color Clear
Material Polystyrene
Well Design Round
Closure Yes
Shelf Life 6 Months
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Accessories

6 Months