Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-L-Lysine Multiwell Plate

Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells

$184.79 - $1,404.97

Specifications

Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
Shelf Life 6 Months
View More Specs
Products
Catalog Number Mfr. No. Quantity Type Price Quantity    

08-772-90

 
corning™
C354515
5/pk. 6-Well Poly-L-Lysine Multiwell Microplate Case of 5 Each for $184.79

08-772-91

 
corning™
C356515
5/pk.; 50/cs. 6-Well Poly-L-Lysine Multiwell Microplate Case of 50 Each for $1,211.85

08-772-92

 
corning™
C354516
5/pk. 96-Well Poly-L-Lysine Multiwell Microplate Pack of 5 Each for $192.62

08-772-93

 
corning™
C356516
5/pk.; 50/cs. 96-Well Poly-L-Lysine Multiwell Microplate Case of 50 Each for $1,404.97
Description & Specifications

Specifications

Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
Shelf Life 6 Months
Source Poly-L-Lysine
Storage Requirements 4° to 30°C
Surface Coating ECM/Attachment Factors
Well Color Clear
Well Design Round

Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.

  • Minimize assay failure due to cell detachment
  • Matrix components are applied as an even, optically clear coating
  • Precoated with Poly-L-Lysine to eliminate time-consuming preparation
  • Lot-to-lot consistency provides reproducible experiments
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
  • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo
  • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes offer convenient viewing of living cells with a phase-contrast microscope
  • Have low-binding properties that minimize small molecule loss

These products are for research use only and are not intended for diagnostic purposes.

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

Safety and Handling

6 Months