Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks


Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Manufacturer: corning™  C356537

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-772-60

  • / Case of 50 Each
 

Description & Specifications

Specifications

Closure Material Phenolic
Shape Rectangular
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
Culture Area 75cm2
Closure Type Vented
Capacity (English) 8.45 oz.
Capacity 8.45 oz., 250mL
Sterile Yes
Capacity (Metric) 250mL
Surface Coating Poly-D-Lysine