Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-D-Lysine Multiwell Plates

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Poly-D-Lysine (PDL) Multiwell Plates are ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

$146.84 - $5,223.59

Specifications

Surface Coating ECM/Attachment Factors
Well Design Round
Material Polystyrene
Bottom Flat
Certifications/Compliance ANSI, SBS
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

 Disclaimers

These products are for research only and not intended for diagnostic purposes.

Products
Catalog Number Mfr. No. Color Format Type Well Color Price Quantity    

08-775-30

 
corning™
C354396
Black 384-well Small volume Clear Case of 5 Each for $403.76

08-775-31

 
corning™
C356396
Black 384-well Small volume Clear Case of 50 Each for $3,182.00

08-774-123

 
corning™
C354413
Clear 6-well Standard Clear Case of 5 Each for $146.84

08-774-127

 
corning™
C354461
Clear 96-well Standard Clear Case of 5 Each for $170.62

08-774-124

 
corning™
C354414
Clear 24-well Standard Clear Case of 5 Each for $162.02

08-774-255

 
corning™
C354640
Black 96-well Standard Clear Case of 5 Each for $313.62

08-774-269

 
corning™
C354470
Clear 12-well Standard Clear Case of 5 Each for $208.10

08-774-270

 
corning™
C356470
Clear 12-well Standard Clear Case of 50 Each for $1,295.51

08-774-280

 
corning™
C354509
Clear 48-well Standard Clear Pack of 5 Each for $289.23

08-774-295

 
corning™
C354620
White 96-well Standard Opaque Case of 5 Each for $237.33

08-774-309

 
corning™
C354660
White 384-well Standard Clear Case of 5 Each for $451.26

08-774-311

 
corning™
C354662
Clear 384-well Standard Clear Case of 5 Each for $257.03

08-774-312

 
corning™
C354663
Black 384-well Standard Clear Case of 5 Each for $441.26

08-774-308

 
corning™
C354651
White 96-well Standard Clear Case of 5 Each for $376.41

08-774-318

 
corning™
C356651
White 96-well Standard Clear Case of 50 Each for $2,590.95

08-774-320

 
corning™
C356661
White 384-well Standard White Case of 50 Each for $2,162.79

08774319

 
corning™
C356660
White 384-well Standard Clear Case of 50 Each for $3,469.33

10-1291

 
corning™
C356690
Clear 96-well Standard Clear Case of 80 Each for $2,024.67

10-1292

 
corning™
C356691
White 96-well Standard White Case of 80 Each for $2,210.44

10-129-3

 
corning™
C356692
Black 96-well Standard Clear Case of 80 Each for $3,717.15

10-1298

 
corning™
C356697
Black 384-well Standard Clear Case of 80 Each for $5,223.59

08-774-271

 
corning™
C356414
Clear 24-well Standard Clear Case of 50 Each for $1,295.51

08-774-256

 
corning™
C356640
Black 96-well Standard Clear Case of 50 Each for $2,467.59

08-774-272

 
corning™
C356461
Clear 96-well Standard Clear Case of 50 Each for $1,535.92

08-774-298

 
corning™
C356620
White 96-well Standard Opaque Case of 50 Each for $1,621.85

08-774-288

 
corning™
C356509
Clear 48-well Standard Clear Case of 50 Each for $1,764.62

08-774-268

 
corning™
C356413
Clear 6-well Standard Clear Case of 50 Each for $1,349.77

08-774-310

 
corning™
C354661
White 384-well Standard White Case of 5 Each for $276.69

08-774-322

 
corning™
C356663
Black 384-well Standard White Case of 50 Each for $3,777.33
Description & Specifications

Specifications

Surface Coating ECM/Attachment Factors
Well Design Round
Material Polystyrene
Bottom Flat
Certifications/Compliance ANSI, SBS
Source Poly-D-Lysine
For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
Closure Yes

 

  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Available in 6, 12, 24, 48, 96 and 384 wells