Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-D-Lysine Multiwell Plates


Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Poly-D-Lysine (PDL) Multiwell Plates are ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Manufacturer: corning™  C354651

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

 Disclaimers

These products are for research only and not intended for diagnostic purposes.

Catalog No. 08-774-308

  • / Case of 5 Each
 

Description & Specifications

Specifications

Bottom Flat
Type Standard
Lid Yes
For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
Format 96-well
Material Polystyrene
Quantity 5/Cs.
Shelf Life 6 Months
Source Poly-D-Lysine
Sterile No
Storage Requirements 4° to 30°C
Surface Coating ECM/Attachment Factors
Certifications/Compliance ANSI, SBS
Well Color Clear
Well Design Round
Closure Yes
Color White
Well Color Clear
Well Shape Round
Bottom Plate Material Polystyrene
Culture Area 0.32cm2
Surface Coating ECM/Attachment Factors
Closure Yes
Applications Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture

 

  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Available in 6, 12, 24, 48, 96 and 384 wells