Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Poly-D-Lysine/Laminin Multiwell Plate

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology

$551.59 - $580.90

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

 Disclaimers

For research use only.

Products
Catalog Number Mfr. No. Culture Area Format Type Price Quantity    

08-774-168

 
corning™
C354595
9.6cm2 6-well 6-Well Poly-D-Lysine/Laminin Multiwell Microplate Case of 5 Each for $551.59

08-774-170

 
corning™
C354596
0.32cm2 96-well 96-Well Poly-D-Lysine/Laminin Multiwell Microplate Case of 5 Each for $580.90

08-774-169

 
corning™
C354619
2cm2 24-well 24-Well Poly-D-Lysine/Laminin Multiwell Microplate Case of 5 Each for $563.44
Description & Specifications

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
Shelf Life 3 Months
Source Poly-D-Lysine and Laminin
Storage Requirements 2° to 8°C
Surface Coating ECM/Attachment Factors
Well Design Round

  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

Applications:

For adhesion, growth and differentiation of neuronal and transfected cell lines

Safety and Handling

3 Months