Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates

Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature

$287.24 - $1,487.10

Specifications

Material Polystyrene
Lid Yes
Color Clear
Sterile No
Shelf Life 3 Months
View More Specs

 Disclaimers

For research use only

Products
Catalog Number Mfr. No. No. of Wells Price Quantity    

08-772-651

 
corning™
C354671
6-well (for ES Culture) Case of 5 Each for $287.24

08-774-108

 
corning™
C354503
12-well Case of 2 Each for $1,176.51

08-774-110

 
corning™
C354508
48-well Case of 2 Each for $1,408.18

08-774-164

 
corning™
C354603
6-well Case of 5 Each for $613.77

08-774-194

 
corning™
C354510
6-well (for hepatocytes) Each for $1,487.10

08-774-165

 
corning™
C354605
24-well Case of 5 Each for $639.54
Description & Specifications

Specifications

Material Polystyrene
Lid Yes
Color Clear
Sterile No
Shelf Life 3 Months
Source EHS Mouse Tumor
Well Design Round

Corning Matrigel Basement Membrane Matrix is a solubilized tissue basement membrane preparation containing laminin, collagen type IV, heparan sulfate proteoglycan and entactin. Corning Matrigel Matrix is useful as a substrate for growth and differentiation of many normal and transformed cell types, such as epithelial, endothelial, muscle and neuronal cells. Corning BioCoat Thin Layer Matrigel cultureware contain a dried thin layer of Corning Matrigel Basement Membrane Matrix. Rehydration results in the formation of a thin “basement membrane” gel layer on the cultureware surface. Specific applications for Thin Layer Matrigel cellware include culturing of primary contractile skeletal myotubes, neuronal cell morphological differentiation, studies of vascular smooth muscle cell interaction with the extracellular matrix and the effects of the extracellular matrix on the function and morphology of lung alveolar, and bile ductular epithelial cells.

  • Especially suited for culture of polarized cells, such as epithelial cells, and promotes differentiation of hepatocytes, mammary epithelial cells and smooth muscle cells
  • Promotes capillary tubule formation of endothelial cells and neurite regeneration
  • Can be used for angiogenesis assays
  • Tested for the ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF)

Safety and Handling

3 Months