Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates


Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature

Manufacturer: corning™  C354605

 View more versions of this product

 Disclaimers

For research use only

Catalog No. 08-774-165

  • / Case of 5 Each
 

Description & Specifications

Specifications

Lid Yes
Closure Yes
Material Polystyrene
Surface Coating ECM/Attachment Factors
Format 24-well
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Source EHS Mouse Tumor
Well Color Clear
Sterile No
Storage Requirements 2° to 8°C
Culture Area 2cm2
No. of Wells 24-well
Well Design Round
Shelf Life 3 Months
Type 24-Well Matrigel Matrix Thin Layer Multiwell Microplate
Color Clear
Accessories

3 Months