Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates
Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature
Manufacturer:  Corning™ C354510
 View more versions of this product
Catalog No. 08-774-194
Disclaimers:
For research use only
Specifications Description & Specifications
Specifications
Yes
9.6cm2
Clear
6-Well Matrigel Matrix Multiwell Microplate
Hepatocytes, elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
No
-20°C
ECM/Attachment Factors
Yes
Polystyrene
6-well (for hepatocytes)
EHS Mouse Tumor
Clear
Round
6-well