Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates


Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature

Manufacturer: corning™  C354508

 View more versions of this product

 Disclaimers

For research use only

Catalog No. 08-774-110

  • / Case of 2 Each
 

Description & Specifications

Specifications

Lid Yes
No. of Wells 48-well
Type 48-Well Matrigel matrix Multiwell Microplate
Shelf Life 3 Months
Storage Requirements -20°C
Culture Area 0.75cm2
Surface Coating ECM/Attachment Factors
Source EHS Mouse Tumor
Well Color Clear
Sterile No
Closure Yes
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Material Polystyrene
Format 48-well
Well Design Round
Color Clear
Accessories

3 Months