Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates


Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature

Manufacturer: corning™  C354503

 View more versions of this product

 Disclaimers

For research use only

Catalog No. 08-774-108

  • / Case of 2 Each
 

Description & Specifications

Specifications

Lid Yes
Format 12-well
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Color Clear
Sterile No
Closure Yes
Source EHS Mouse Tumor
Culture Area 3.8cm2
Shelf Life 3 Months
Storage Requirements -20°C
Well Color Clear
Material Polystyrene
Type 12-Well Matrigel Matrix Thin Layer Multiwell Microplate
No. of Wells 12-well
Surface Coating ECM/Attachment Factors
Well Design Round
Accessories

3 Months