Learn More
Corning™ BioCoat™ Laminin TC-treated Culture Dishes
Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
$557.36 - $777.58
Specifications
Bottom Shape | Round |
---|---|
Color | Clear |
Material | Polystyrene |
Shape | Round |
Type | Cell Culture Dish |
Description
Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.
Applications include:
- Promotion of cell attachment and spreading
- Induction of cell differentiation and neurite outgrowth
- Increases proliferation of myoblasts
- Studies of effects of laminin on cell behavior
- Cell adhesion assays
Source
Quality Control
- Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
- Tested and found negative for bacteria and fungi
- Laminin purity >90% by SDS-PAGE (contains entactin)
Storage
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Round | |
Polystyrene | |
Cell Culture Dish |
Clear | |
Round | |
Laminin |