Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap


Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

Manufacturer: corning™  C354533

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-93

  • / Case of 10 Each
 

Description & Specifications

Specifications

Capacity 2.36 oz., 70mL
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Closure Material Phenolic
Sterile Yes
Shape Rectangular
Capacity (Metric) 70mL
Neck Style Canted
Culture Area 25cm2
Surface Coating Laminin
Neck Finish Canted
Capacity (English) 2.36 oz.