Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Laminin Multiwell Plate


Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

Manufacturer: corning™  C354507

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-85

  • / Case of 5 Each
 

Description & Specifications

Specifications

Lid Yes
Culture Area 0.75cm2
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Format 48-well
Material Polystyrene
Shelf Life 3 Months
Source Mouse Laminin
Sterile No
Storage Requirements 2° to 8°C
Surface Coating ECM/Attachment Factors
Type 48-Well Laminin Multiwell Microplate
Well Color Clear
Well Design Round
Closure Yes
Color Clear
Applications Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Surface Coating ECM/Attachment Factors
Well Shape Round
Bottom Plate Material Polystyrene
Well Color Clear
Closure Yes