Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Laminin Multiwell Plate

Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

$437.59 - $577.05

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Culture Area Format Type Price Quantity    

08-774-83

 
corning™
C354502
3.8cm2 12-well 12-Well Laminin Multiwell Microplate Case of 5 Each for $504.08

08-774-82

 
corning™
C354404
9.6cm2 6-well 6-Well Laminin Multiwell Microplate Case of 5 Each for $437.59

08-774-84

 
corning™
C354412
2cm2 24-well 24-Well Laminin Multiwell Microplate Case of 5 Each for $445.49

08-774-85

 
corning™
C354507
0.75cm2 48-well 48-Well Laminin Multiwell Microplate Case of 5 Each for $546.33

08-774-86

 
corning™
C354410
0.32cm2 96-well 96-Well Laminin Multiwell Microplate Case of 5 Each for $577.05
Description & Specifications

Specifications

Material Polystyrene
Lid Yes
For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Color Clear
Sterile No
Shelf Life 3 Months
Source Mouse Laminin
Storage Requirements 2° to 8°C
Surface Coating ECM/Attachment Factors
Well Design Round

  • Designed to closely mimic in vivo environments
  • Matrix components are applied as an even, optically clear coating

These products are for research use only and are not intended for diagnostic purposes.

Safety and Handling

3 Months