Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Gelatin Rectangular Canted Neck Culture Flasks w/Vented Cap


Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation

Manufacturer: corning™  C354488

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-331

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Shape Rectangular
Closure Material Phenolic
No. per Pack 5.0
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
Capacity (English) 8.45 oz.
Capacity 8.45 oz., 250mL
Capacity (Metric) 250mL
Culture Area 75cm2
Sterile Yes
Surface Coating Gelatin
Neck Finish Straight