Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Fibronectin Rectangular Canted Neck Culture Flask


Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells

Manufacturer: corning™  C354532

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-68

  • / Case of 10 Each
 

Description & Specifications

Specifications

For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Shape Rectangular
Culture Area 25cm2
Closure Type Plug-seal
Capacity (English) 2.36 oz.
Capacity 2.36 oz., 70mL
Closure Material Phenolic
Sterile Yes
Capacity (Metric) 70mL
Surface Coating Fibronectin
Neck Style Canted
Neck Finish Straight