missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Fibronectin Multiwell Plate

Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells

$417.78 - $1,339.05

Specifications

Lid Yes
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
Sterile No
Source Human Fibronectin
Storage Requirements 2° to 8°C
View More Specs

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.



Products ${productFamilyLength}
Catalog Number Mfr. No. Color Format Type Well Color Price Quantity  
Catalog Number Mfr. No. Color Format Type Well Color Price Quantity  
08-774-56 Corning™
C354402
Clear 6-well 6-Well Fibronectin Multiwell Microplate Clear Case of 5 Each for $417.78
08-774-58 Corning™
C354411
Clear 24-well 24-Well Fibronectin Multiwell Microplate Clear Case of 5 Each for $427.69
08-774-57 Corning™
C354501
Clear 12-well 12-Well Fibronectin Multiwell Microplate Clear Case of 5 Each for $484.54
08-774-59 Corning™
C354506
Clear 48-well 48-Well Fibronectin Multiwell Microplate Clear Case of 5 Each for $564.13
08-774-60 Corning™
C354409
Clear 96-well 96-Well Fibronectin Multiwell Microplate Clear Case of 5 Each for $486.31
15-100-162 Corning™
C4584
Half Area Black/Clear 96-well 96-Well Fibronectin Multiwell Microplate Half Area Black/Clear Case for $1,339.05
Specifications

Description

  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
  • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo
  • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes offer convenient viewing of living cells with a phase-contrast microscope
  • Have low-binding properties that minimize small molecule loss

The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody. These products are for research use only and are not intended for diagnostic purposes.

Order Info

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

Description & Specifications

Specifications

Yes
No
2° to 8°C
Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
Human Fibronectin
Round