Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Fibronectin, Human Cell Culture Insert

Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells

$909.77 - $929.72

Specifications

Material PET
Type Membrane Insert
For Use With (Equipment) 24 Well Plate
Storage Requirements 2° to 8°C
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Pore Size Price Quantity    

08-774-74

 
corning™
C354445
0.4μm Case of 24 Each for $929.72

08-774-76

 
corning™
C354543
3μm Case of 24 Each for $909.77
Description & Specifications

Specifications

Material PET
Type Membrane Insert
For Use With (Equipment) 24 Well Plate
Storage Requirements 2° to 8°C

  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
  • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces
  • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes for convenient viewing of living cells with a phase-contrast microscope
  • Have low-binding properties that minimize small molecule loss

The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates