Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Endothelial Cell Growth Environment Kit


Increases endothelial cell yield from a variety of sources

Manufacturer: corning™  C355053

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-149

  • / Each
 

Description & Specifications

Specifications

Type Complete Endothelial Cell Growth Environment Kit
For Use With (Application) Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Storage Requirements 2° to 8°C