Please login to your online account to display your discounted pricing
Corning™ BioCoat™ Control Insert
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Manufacturer: corning™  C354576
 View more versions of this product
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog No. 08-774-161
  • / Case of 24 Each
Description & Specifications
Specifications
Inserts per Plate 6
Quantity 4
No. of Wells 6
Type Well Plate
Storage Requirements 2° to 8°C
Pore Size 8μm