Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask


Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Manufacturer: corning™  C354534

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-37

  • / Case of 10 Each
 

Description & Specifications

Specifications

Closure Type Plug-seal
Closure Material Phenolic
Shape Rectangular
Neck Style Canted
Culture Area 25cm2
Sterile Yes
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Capacity (Metric) 70mL
Surface Coating Collagen IV

 

  • BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

 

For research use only.