Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Collagen IV Multiwell Plates

Tissue culture vessels have a uniform application of mouse collagen type IV

$446.00 - $587.79

Specifications

Material Polystyrene
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Color Clear
Sterile No
Shelf Life 3 Months
View More Specs

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Products
Catalog Number Mfr. No. Format Type Price Quantity    

08-774-28

 
corning™
C354428
6-well 6-Well Collagen IV Multiwell Microplate Case of 5 Each for $446.00

08-774-29

 
corning™
C354430
24-well 24-Well Collagen IV Multiwell Microplate Case of 5 Each for $454.14

08-774-30

 
corning™
C354429
96-well 96-Well Collagen IV Multiwell Microplate Case of 5 Each for $587.79
Description & Specifications

Specifications

Material Polystyrene
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Color Clear
Sterile No
Shelf Life 3 Months
Source EHS Lathrytic Mouse Tumor
Storage Requirements 2° to 8°C
Surface Coating ECM/Attachment Factors
Well Design Round

Treated with Collagen IV, which plays a role in the regulation of cell growth, differentiation, and adhesion, as well as tissue formation.

  • Tested for the ability to promote differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
  • Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance

For research use only—not intended for diagnostic purposes