Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap


Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.

Manufacturer: corning™  C354484

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-327

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Closure Material Phenolic
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Shape Rectangular
Culture Area 25cm2
Capacity (Metric) 70mL
Surface Coating Collagen I
Capacity (English) 2.36 oz.
Capacity 2.36 oz., 70mL
No. per Case 10/Cs.
Sterile Yes
Neck Style Straight
Closure Type Vented
Barcode Yes

  • Vented caps allow gas exchange and minimize risk of contamination
  • Ideal for adhesion and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.